Alfa Aesar™ 2,4-Dichloro-5-fluorobenzoyl chloride, 97%

Manufacturer: Alfa Aesar™ B2351403

Quantity 1g

 View more versions of this product

Catalog No. AAB2351403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 86393-34-2
    Molecular Formula C7H2Cl3FO
    Formula Weight 227.45
    MDL Number MFCD00075341
    UN Number UN3265
    Beilstein 4989077
    Chemical Name or Material 2,4-Dichloro-5-fluorobenzoyl chloride

    Specifications

    Formula Weight 227.45
    UN Number UN3265
    MDL Number MFCD00075341
    CAS 86393-34-2
    Beilstein 4989077
    Flash Point >110°C (230°F)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H317-H335
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Quantity 1g
    Molecular Formula C7H2Cl3FO
    Boiling Point 143° to 144°C (35mmHg)
    Health Hazard 3 GHS P Statement
    P280-P262-P273-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    Do not get in eyes, on skin, or on clothing.
    Avoid release to the environment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Chemical Name or Material 2,4-Dichloro-5-fluorobenzoyl chloride
    Assay 97%

    GHS H Statement
    H314-H318-H317-H335
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.
    May cause respiratory irritation.

    GHS P Statement
    P280-P262-P273-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    Do not get in eyes, on skin, or on clothing.
    Avoid release to the environment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit