Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Dichloro-5-fluorobenzoyl chloride, 97%

Click to view available options
Quantity:
1g
5g
86393-34-2
C7H2Cl3FO
227.44
MFCD00075341
RPZXUSJCSDQNTE-UHFFFAOYSA-N
2,4-dichloro-5-fluoro-benzoyl chloride,2,4-dichloro-5-fluoro benzoyl chloride,benzoyl chloride,2,4-dichloro-5-fluoro,2,4-dichloro-5-fluorobenzoylchloride,2,4-bis chloranyl-5-fluoranyl-benzoyl chloride
2736821
2,4-dichloro-5-fluorobenzoyl chloride
C1=C(C(=CC(=C1F)Cl)Cl)C(=O)Cl
86393-34-2
C7H2Cl3FO
227.44
MFCD00075341
RPZXUSJCSDQNTE-UHFFFAOYSA-N
2,4-dichloro-5-fluoro-benzoyl chloride,2,4-dichloro-5-fluoro benzoyl chloride,benzoyl chloride,2,4-dichloro-5-fluoro,2,4-dichloro-5-fluorobenzoylchloride,2,4-bis chloranyl-5-fluoranyl-benzoyl chloride
2736821
2,4-dichloro-5-fluorobenzoyl chloride
C1=C(C(=CC(=C1F)Cl)Cl)C(=O)Cl
Boiling Point 143°C to 144°C (35mmHg)
Quantity 5g
Flash Point >110°C (230°F)
UN Number UN3265
Beilstein 4989077
Formula Weight 227.45
Percent Purity 97%
Chemical Name or Material 2,4-Dichloro-5-fluorobenzoyl chloride

GHS H Statement
H314-H318-H317-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P280-P262-P273-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.