Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-5-fluoropyrimidine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
2927-71-1
C4HCl2FN2
166.97
WHPFEQUEHBULBW-UHFFFAOYSA-N
2,4-dichloro-5-fluoro-pyrimidine,5-fluoro-2,4-dichloropyrimidine,2,6-dichloro-5-fluororacil,2,4-dichloro-5-fluoro pyrimidine,pyrimidine, 2,4-dichloro-5-fluoro,2,6-dichloro-5-fluoropyrimidine,2,4-dichlor-5-fluorpyrimidin,pubchem5290,ksc204s2l,2,4-dichloro-5fluoropyrimidine
250705
2,4-dichloro-5-fluoropyrimidine
C1=C(C(=NC(=N1)Cl)Cl)F
2927-71-1
C4HCl2FN2
166.97
WHPFEQUEHBULBW-UHFFFAOYSA-N
2,4-dichloro-5-fluoro-pyrimidine,5-fluoro-2,4-dichloropyrimidine,2,6-dichloro-5-fluororacil,2,4-dichloro-5-fluoro pyrimidine,pyrimidine, 2,4-dichloro-5-fluoro,2,6-dichloro-5-fluoropyrimidine,2,4-dichlor-5-fluorpyrimidin,pubchem5290,ksc204s2l,2,4-dichloro-5fluoropyrimidine
250705
2,4-dichloro-5-fluoropyrimidine
C1=C(C(=NC(=N1)Cl)Cl)F
Melting Point 38°C to 40°C
Boiling Point 67°C to 68°C (6.0 Torr)
Packaging Glass Bottle
Infrared Spectrum Authentic
Quantity 5g
Color Brown to White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
UN Number 3261
Formula Weight 166.97
Physical Form Powder or Crystalline Powder or Low Melting Solid
Percent Purity 97%
Chemical Name or Material 2, 4-Dichloro-5-fluoropyrimidine
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger