Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Dichloro-5-methoxyaniline, 98%

Click to view available options
Quantity:
1g
5g
25g
98446-49-2
C7H7Cl2NO
192.039
MFCD00974410
AJROJTARXSATEB-UHFFFAOYSA-N
5-amino-2,4-dichloroanisole,2,4-dichloro-5-methoxybenzenamine,4,6-dichloro-m-anisidine,2,4-dichloro-5-methoxy-phenylamine,benzenamine, 2,4-dichloro-5-methoxy,2,4-dichloro-5-methoxyphenylamine,dichloromethoxyaniline,pubchem4393,acmc-209sae,4,5-dichloro-m-anisidine
1476636
2,4-dichloro-5-methoxyaniline
COC1=C(C=C(C(=C1)N)Cl)Cl
98446-49-2
C7H7Cl2NO
192.039
MFCD00974410
AJROJTARXSATEB-UHFFFAOYSA-N
5-amino-2,4-dichloroanisole,2,4-dichloro-5-methoxybenzenamine,4,6-dichloro-m-anisidine,2,4-dichloro-5-methoxy-phenylamine,benzenamine, 2,4-dichloro-5-methoxy,2,4-dichloro-5-methoxyphenylamine,dichloromethoxyaniline,pubchem4393,acmc-209sae,4,5-dichloro-m-anisidine
1476636
2,4-dichloro-5-methoxyaniline
COC1=C(C=C(C(=C1)N)Cl)Cl
Quantity 25g
UN Number UN2810
Formula Weight 192.05
Percent Purity 98%
Chemical Name or Material 2,4-Dichloro-5-methoxyaniline

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.