Alfa Aesar™ 2,4-Dichloro-5-methylpyrimidine, 98%

Manufacturer: Alfa Aesar™ H5520606

Quantity 5g

 View more versions of this product

Catalog No. AAH5520606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1780-31-0
    Molecular Formula C5H4Cl2N2
    Formula Weight 163.01
    MDL Number MFCD00023197
    UN Number UN3261
    Chemical Name or Material 2,4-Dichloro-5-methylpyrimidine

    Specifications

    Formula Weight 163.01
    TSCA TSCA
    UN Number UN3261
    Boiling Point 108° to 109°C (11mmHg)
    Melting Point 26° to 28°C
    Quantity 5g
    Molecular Formula C5H4Cl2N2
    MDL Number MFCD00023197
    CAS 1780-31-0
    Flash Point 113°C (235°F)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material 2,4-Dichloro-5-methylpyrimidine
    Assay 98%

    Description

    Applications
    2,4-Dichloro-5-methylpyrimidine, is used in the synthesis of (2-chloro-6-methyl-pyrimidin-4-yl)-(2,3-dihydro-benzothiazol-6-yl)-amine. It can react with piperidineto produce 2-chloro-5-methyl-4-piperidin-1-yl-pyrimidine. This reaction will need solvent dioxane.

    Solubility
    Soluble in chloroform, ether, ethyl acetate and toluene

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit