Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Dichloro-5-nitrobenzotrifluoride, 97%

Click to view available options
Quantity:
5g
25g
100g
400-70-4
C7H2Cl2F3NO2
259.993
MFCD00007073
VLVNHMVSVDVAOA-UHFFFAOYSA-N
2,4-dichloro-5-nitrobenzotrifluoride,1,5-dichloro-2-nitro-4-trifluoromethyl benzene,2,4-dichloro-3,5-nitrobenzotrifluoride,benzene, 1,5-dichloro-2-nitro-4-trifluoromethyl,2,4-dichloro-5-trifluoromethyl-1-nitrobenzene,pubchem4862,2,4-dichloro-5-nitro benzotrifluoride,3-trifluoromethyl-4,6-dichloronitrobenzene,2,4-dichloro-5-trifluoromethyl nitrobenzene
621179
1,5-dichloro-2-nitro-4-(trifluoromethyl)benzene
C1=C(C(=CC(=C1[N+](=O)[O-])Cl)Cl)C(F)(F)F
400-70-4
C7H2Cl2F3NO2
259.993
MFCD00007073
VLVNHMVSVDVAOA-UHFFFAOYSA-N
2,4-dichloro-5-nitrobenzotrifluoride,1,5-dichloro-2-nitro-4-trifluoromethyl benzene,2,4-dichloro-3,5-nitrobenzotrifluoride,benzene, 1,5-dichloro-2-nitro-4-trifluoromethyl,2,4-dichloro-5-trifluoromethyl-1-nitrobenzene,pubchem4862,2,4-dichloro-5-nitro benzotrifluoride,3-trifluoromethyl-4,6-dichloronitrobenzene,2,4-dichloro-5-trifluoromethyl nitrobenzene
621179
1,5-dichloro-2-nitro-4-(trifluoromethyl)benzene
C1=C(C(=CC(=C1[N+](=O)[O-])Cl)Cl)C(F)(F)F
Melting Point 53°C to 57°C
Quantity 5g
Flash Point >110°C (230°F)
Beilstein 2467459
Formula Weight 260
Percent Purity 97%
Chemical Name or Material 2,4-Dichloro-5-nitrobenzotrifluoride

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures