Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-5-sulfamoylbenzoic acid, 98%, Thermo Scientific Chemicals

Catalog Number AAB2458918
Click to view available options
Quantity:
50 g
2736-23-4
C7H5Cl2NO4S
270.08
MFCD00007931
ZSHHRBYVHTVRFK-UHFFFAOYSA-N
lasamide, 5-aminosulfonyl-2,4-dichlorobenzoic acid, 2,4-dichloro-5-sulphamoylbenzoic acid, 2,4-dichloro-5-sulfamoyl-benzoic acid, unii-leg53tf0sn, 2,4-dichloro-5-sulfamylbenzoic acid, benzoic acid, 5-aminosulfonyl-2,4-dichloro, leg53tf0sn, 3-sulfamoyl-4,6-dichlorobenzoic acid, benzoic acid, 2,4-dichloro-5-sulfamoyl
17655
2,4-dichloro-5-sulfamoylbenzoic acid
C1=C(C(=CC(=C1S(=O)(=O)N)Cl)Cl)C(=O)O
2736-23-4
C7H5Cl2NO4S
270.08
MFCD00007931
ZSHHRBYVHTVRFK-UHFFFAOYSA-N
lasamide, 5-aminosulfonyl-2,4-dichlorobenzoic acid, 2,4-dichloro-5-sulphamoylbenzoic acid, 2,4-dichloro-5-sulfamoyl-benzoic acid, unii-leg53tf0sn, 2,4-dichloro-5-sulfamylbenzoic acid, benzoic acid, 5-aminosulfonyl-2,4-dichloro, leg53tf0sn, 3-sulfamoyl-4,6-dichlorobenzoic acid, benzoic acid, 2,4-dichloro-5-sulfamoyl
17655
2,4-dichloro-5-sulfamoylbenzoic acid
C1=C(C(=CC(=C1S(=O)(=O)N)Cl)Cl)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 230°C to 232°C (decomposition)
Quantity 50 g
Formula Weight 270.09
Percent Purity 98%
Chemical Name or Material 2,4-Dichloro-5-sulfamoylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-358-3
RTECSNumber DG8100000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit