Alfa Aesar™ 2,4-Dichloro-5-sulfamoylbenzoic acid, 98%

Manufacturer: Alfa Aesar™ B2458930

Quantity 250g

 View more versions of this product

Catalog No. AAB2458930

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2736-23-4
    Molecular Formula C7H5Cl2NO4S
    Formula Weight 270.09
    MDL Number MFCD00007931
    Chemical Name or Material 2,4-Dichloro-5-sulfamoylbenzoic acid

    Specifications

    Formula Weight 270.09
    Quantity 250g
    Molecular Formula C7H5Cl2NO4S
    MDL Number MFCD00007931
    CAS 2736-23-4
    Melting Point 230° to 232°C (dec.)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H318-H335-H315
    Causes serious eye damage.
    May cause respiratory irritation.
    Causes skin irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number DG8100000
    Chemical Name or Material 2,4-Dichloro-5-sulfamoylbenzoic acid
    Assay 98%

    GHS H Statement
    H318-H335-H315
    Causes serious eye damage.
    May cause respiratory irritation.
    Causes skin irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit