Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-5-(trifluoromethyl)pyrimidine, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
3932-97-6
C5HCl2F3N2
216.98
IDRUEHMBFUJKAK-UHFFFAOYSA-N
2,4-dichloro-5-trifluoromethyl pyrimidine,2,4-dichloro-5-trifluoromethyl-pyrimidine,5-trifluoromethyl-2,4-dichloropyrimidine,pyrimidine, 2,4-dichloro-5-trifluoromethyl,pubchem5304,acmc-1cn5t,c5hcl12f3n2,2,4-dichloro-5-trifluoromethylpyrimidin,2,6-dichloro-5-trifluoromethylpyrimidine,2.4-dichloro-5-trifluoromethylpyrimidine
2782774
2,4-dichloro-5-(trifluoromethyl)pyrimidine
C1=C(C(=NC(=N1)Cl)Cl)C(F)(F)F
3932-97-6
C5HCl2F3N2
216.98
IDRUEHMBFUJKAK-UHFFFAOYSA-N
2,4-dichloro-5-trifluoromethyl pyrimidine,2,4-dichloro-5-trifluoromethyl-pyrimidine,5-trifluoromethyl-2,4-dichloropyrimidine,pyrimidine, 2,4-dichloro-5-trifluoromethyl,pubchem5304,acmc-1cn5t,c5hcl12f3n2,2,4-dichloro-5-trifluoromethylpyrimidin,2,6-dichloro-5-trifluoromethylpyrimidine,2.4-dichloro-5-trifluoromethylpyrimidine
2782774
2,4-dichloro-5-(trifluoromethyl)pyrimidine
C1=C(C(=NC(=N1)Cl)Cl)C(F)(F)F
Specific Gravity 1.609
Density 1.6090g/mL
Quantity 5g
Flash Point 93°C
Boiling Point 48.0°C to 50.0°C (32.0mmHg)
Assay Percent Range 97%
Formula Weight 216.98
Percent Purity 97%
Chemical Name or Material 2,4-Dichloro-5-(trifluoromethyl)pyrimidine

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYE

GHS Signal Word: Danger