Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-6,7-dimethoxyquinazoline, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
27631-29-4
C10H8Cl2N2O2
259.09
MFCD00051733
DGHKCBSVAZXEPP-UHFFFAOYSA-N
2,4-dichloro-6,7-dimethoxy-quinazoline,2,4-dichloro-6,7-dimethoxy quinazoline,quinazoline, 2,4-dichloro-6,7-dimethoxy,2-chloro-6,7-dimethoxyquinazolin-4-yl chloride,quinazoline,2,4-dichloro-6,7-dimethoxy,doxazosin impurity e,doxazosin mesilate impurity e ep,pubchem9430,usp doxazosin related compound e usp,acmc-209gyd
520327
2,4-dichloro-6,7-dimethoxyquinazoline
COC1=C(C=C2C(=C1)C(=NC(=N2)Cl)Cl)OC
27631-29-4
C10H8Cl2N2O2
259.09
MFCD00051733
DGHKCBSVAZXEPP-UHFFFAOYSA-N
2,4-dichloro-6,7-dimethoxy-quinazoline,2,4-dichloro-6,7-dimethoxy quinazoline,quinazoline, 2,4-dichloro-6,7-dimethoxy,2-chloro-6,7-dimethoxyquinazolin-4-yl chloride,quinazoline,2,4-dichloro-6,7-dimethoxy,doxazosin impurity e,doxazosin mesilate impurity e ep,pubchem9430,usp doxazosin related compound e usp,acmc-209gyd
520327
2,4-dichloro-6,7-dimethoxyquinazoline
COC1=C(C=C2C(=C1)C(=NC(=N2)Cl)Cl)OC
Infrared Spectrum Authentic
Melting Point 172°C to 177°C
Color Beige to Yellow
Quantity 5g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 259.09
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 2, 4-Dichloro-6, 7-dimethoxyquinazoline, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA