Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Dichloro-6-(trifluoromethyl)pyrimidine , 95%

Click to view available options
Quantity:
250mg
1g
5g
16097-64-6
C5HCl2F3N2
216.972
MFCD09910264
ZTNFYAJHLPMNSN-UHFFFAOYSA-N
2,4-dichloro-6-trifluoromethyl pyrimidine,pyrimidine, 2,4-dichloro-6-trifluoromethyl,2,4-dichloro-6-trifluoromethyl-pyrimidine,pubchem13247,ksc496a7p,2,4-dichloro-6-trifluoromethyl-pyrimidine,2,6-dichloro-4-trifluoromethyl pyrimidine
334203
2,4-dichloro-6-(trifluoromethyl)pyrimidine
C1=C(N=C(N=C1Cl)Cl)C(F)(F)F
16097-64-6
C5HCl2F3N2
216.972
MFCD09910264
ZTNFYAJHLPMNSN-UHFFFAOYSA-N
2,4-dichloro-6-trifluoromethyl pyrimidine,pyrimidine, 2,4-dichloro-6-trifluoromethyl,2,4-dichloro-6-trifluoromethyl-pyrimidine,pubchem13247,ksc496a7p,2,4-dichloro-6-trifluoromethyl-pyrimidine,2,6-dichloro-4-trifluoromethyl pyrimidine
334203
2,4-dichloro-6-(trifluoromethyl)pyrimidine
C1=C(N=C(N=C1Cl)Cl)C(F)(F)F
Refractive Index 1.4640
Color Yellow
Quantity 250mg
Density 1.589g/mL
Beilstein 911050
Formula Weight 216.97
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 2,4-Dichloro-6-(trifluoromethyl)pyrimidine

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning