Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-7H-pyrrolo[2,3-d]pyrimidine, 95%, Thermo Scientific™

Click to view available options
Quantity:
250mg
1g
90213-66-4
C6H3Cl2N3
188.02
GHXBPCSSQOKKGB-UHFFFAOYSA-N
2,4-dichloro-7h-pyrrolo 2,3-d pyrimidine, 2,4-dichloro-1h-pyrrolo 2,3-d pyrimidine, 2,6-dichloro-7-deazapurine, 2,4-dichloropyrrolo 2,3-d pyrimidine, 2,4-dichloro-7h-pyrrolo2,3-dpyrimidine, 1h-pyrrolo 2,3-d pyrimidine, 2,4-dichloro, pubchem14941, acmc-209unt, tofacitinib intermediate ii, ksc486m2l
14116871
2,4-dichloro-7H-pyrrolo[2,3-d]pyrimidine
C1=CNC2=C1C(=NC(=N2)Cl)Cl
90213-66-4
C6H3Cl2N3
188.02
GHXBPCSSQOKKGB-UHFFFAOYSA-N
2,4-dichloro-7h-pyrrolo 2,3-d pyrimidine, 2,4-dichloro-1h-pyrrolo 2,3-d pyrimidine, 2,6-dichloro-7-deazapurine, 2,4-dichloropyrrolo 2,3-d pyrimidine, 2,4-dichloro-7h-pyrrolo2,3-dpyrimidine, 1h-pyrrolo 2,3-d pyrimidine, 2,4-dichloro, pubchem14941, acmc-209unt, tofacitinib intermediate ii, ksc486m2l
14116871
2,4-dichloro-7H-pyrrolo[2,3-d]pyrimidine
C1=CNC2=C1C(=NC(=N2)Cl)Cl
Quantity 1g
Packaging Glass Bottle
Melting Point 250°C to 252°C
Assay Percent Range 94% min. (HPLC)
Formula Weight 188.02
Percent Purity 95%
Chemical Name or Material 2, 4-Dichloro-7H-pyrrolo[2, 3-d]pyrimidine

GHS Signal Word: Warning

RUO – Research Use Only