Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-α-methylbenzyl alcohol, 99+%, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
1475-13-4
C8H8Cl2O
191.06
MFCD00004511
KWZDYNBHZMQRLS-UHFFFAOYSA-N
1-2,4-dichlorophenyl ethanol,1-2,4-dichlorophenyl ethan-1-ol,2,4-dichloro-alpha-methylbenzyl alcohol,1-2,4-dichloro-phenyl ethanol,1-2,4-dichlorophenyl ethyl alcohol,2,4-dichloro-alpha-methylbenzylalcohol,2,4-dichloro-1-1-hydroxyethyl benzene,2,4-dichloro-alpha-methylbenzenemethanol,2,4-dichloro-.alpha.-methylbenzyl alcohol,2,4-dichloro-a-methylbenzyl alcohol
92889
1-(2,4-dichlorophenyl)ethanol
CC(C1=C(C=C(C=C1)Cl)Cl)O
1475-13-4
C8H8Cl2O
191.06
MFCD00004511
KWZDYNBHZMQRLS-UHFFFAOYSA-N
1-2,4-dichlorophenyl ethanol,1-2,4-dichlorophenyl ethan-1-ol,2,4-dichloro-alpha-methylbenzyl alcohol,1-2,4-dichloro-phenyl ethanol,1-2,4-dichlorophenyl ethyl alcohol,2,4-dichloro-alpha-methylbenzylalcohol,2,4-dichloro-1-1-hydroxyethyl benzene,2,4-dichloro-alpha-methylbenzenemethanol,2,4-dichloro-.alpha.-methylbenzyl alcohol,2,4-dichloro-a-methylbenzyl alcohol
92889
1-(2,4-dichlorophenyl)ethanol
CC(C1=C(C=C(C=C1)Cl)Cl)O
Specific Gravity 1.29
Flash Point 108°C
Packaging Glass bottle
Color Colorless
Density 1.2900g/mL
Quantity 10g
Infrared Spectrum Authentic
Refractive Index 1.5590 to 1.5610
Boiling Point 125.0°C to 126.0°C (7.0 mmHg)
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula Cl2C6H3CH(CH3)OH
Beilstein 06, II, 446
Formula Weight 191.06
Physical Form Liquid
Percent Purity 99+%
Chemical Name or Material 2,4-Dichloro-α-methylbenzyl alcohol, 99+%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 216-019-4

TSCA : TSCA