Alfa Aesar™ 2',4'-Dichloroacetanilide, 98%

Manufacturer: Alfa Aesar™ B2179588

Quantity 2.5g

 View more versions of this product

Catalog No. AAB2179588

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 6975-29-7
    Molecular Formula C8H7Cl2NO
    Formula Weight 204.06
    MDL Number MFCD00045181
    Chemical Name or Material 2',4'-Dichloroacetanilide

    Specifications

    Formula Weight 204.06
    Melting Point 146° to 148°C
    Quantity 2.5g
    TSCA TSCA
    Molecular Formula C8H7Cl2NO
    MDL Number MFCD00045181
    CAS 6975-29-7
    RTECS Number AE1945000
    Chemical Name or Material 2',4'-Dichloroacetanilide
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit