Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',4'-Dichloroacetophenone, 94%, ACROS Organics™

Click to view available options
Quantity:
100g
250g
500g
2234-16-4
C8H6Cl2O
189.035
XMCRWEBERCXJCH-UHFFFAOYSA-N
2',4'-dichloroacetophenone,1-2,4-dichlorophenyl ethanone,2,4-dichloroacetophenone,ethanone, 1-2,4-dichlorophenyl,1-2,4-dichlorophenyl ethan-1-one,p-chloro-2-chloroacetophenone,acetophenone, 2',4'-dichloro,unii-t7f2608h5h,2',4'-dichloro phenyl ethanone,1-2,4-dichloro-phenyl-ethanone
16693
1-(2,4-dichlorophenyl)ethanone
CC(=O)C1=C(C=C(C=C1)Cl)Cl
2234-16-4
C8H6Cl2O
189.035
XMCRWEBERCXJCH-UHFFFAOYSA-N
2',4'-dichloroacetophenone,1-2,4-dichlorophenyl ethanone,2,4-dichloroacetophenone,ethanone, 1-2,4-dichlorophenyl,1-2,4-dichlorophenyl ethan-1-one,p-chloro-2-chloroacetophenone,acetophenone, 2',4'-dichloro,unii-t7f2608h5h,2',4'-dichloro phenyl ethanone,1-2,4-dichloro-phenyl-ethanone
16693
1-(2,4-dichlorophenyl)ethanone
CC(=O)C1=C(C=C(C=C1)Cl)Cl
Quantity 250g
Density 1.32g/mL
CAS Min % 92.0
CAS Max % 100.0
Assay Percent Range 94%
Linear Formula Cl2C6H3COCH3
Beilstein 07, II, 219
Formula Weight 189.04
Chemical Name or Material 2', 4'-Dichloroacetophenone, 94%

GHS H Statement:
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 218-780-8

TSCA : TSCA