Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloroaniline, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
250g
1kg
2.5kg
Packaging:
Glass bottle
Plastic bottle
554-00-7
C6H5Cl2N
162.02
MFCD00007661
KQCMTOWTPBNWDB-UHFFFAOYSA-N
2,4-dichlorobenzenamine,benzenamine, 2,4-dichloro,2,4-dichloranilin,o,p-dichloroaniline,aniline, 2,4-dichloro,2,4-dca,2,4-dichlorophenylamine,2,4-dichloro-phenylamine,unii-19ae42m6ws,2,4-dichloranilin german
11123
CHEBI:46635
2,4-dichloroaniline
C1=CC(=C(C=C1Cl)Cl)N
554-00-7
C6H5Cl2N
162.02
MFCD00007661
KQCMTOWTPBNWDB-UHFFFAOYSA-N
2,4-dichlorobenzenamine,benzenamine, 2,4-dichloro,2,4-dichloranilin,o,p-dichloroaniline,aniline, 2,4-dichloro,2,4-dca,2,4-dichlorophenylamine,2,4-dichloro-phenylamine,unii-19ae42m6ws,2,4-dichloranilin german
11123
CHEBI:46635
2,4-dichloroaniline
C1=CC(=C(C=C1Cl)Cl)N
Boiling Point 242°C
Flash Point >110°C
Packaging Plastic bottle
Quantity 2.5kg
Infrared Spectrum Authentic
Color White to Beige
Melting Point 59°C to 63°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula Cl2C6H3NH2
Beilstein 12,621
Solubility Information Solubility in water: insoluble
Formula Weight 162.02
Physical Form Crystalline Powder
Percent Purity ≥97.5% (GC)
Chemical Name or Material 2, 4-Dichloroaniline, 98%
Show More Show Less

GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-057-8

RTECSNumber : BX2600000

TSCA : TSCA

Recommended Storage : Product darkens on storage