Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorobenzamide, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
2447-79-2
C7H5Cl2NO
190.023
MFCD00007974
XXFUNTSOBHSMBU-UHFFFAOYSA-N
benzamide, 2,4-dichloro,unii-zx274bu5ra,zx274bu5ra,2,4-dichloro-benzamide,acmc-1crzq,xxfuntsobhsmbu-uhfffaoysa
75556
2,4-dichlorobenzamide
C1=CC(=C(C=C1Cl)Cl)C(=O)N
2447-79-2
C7H5Cl2NO
190.023
MFCD00007974
XXFUNTSOBHSMBU-UHFFFAOYSA-N
benzamide, 2,4-dichloro,unii-zx274bu5ra,zx274bu5ra,2,4-dichloro-benzamide,acmc-1crzq,xxfuntsobhsmbu-uhfffaoysa
75556
2,4-dichlorobenzamide
C1=CC(=C(C=C1Cl)Cl)C(=O)N
Melting Point 191°C to 194°C
Infrared Spectrum Authentic
Quantity 25g
Color White
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula Cl2C6H3CONH2
Beilstein 09, III, 1376
Formula Weight 190.03
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2, 4-Dichlorobenzamide, 98%
Show More Show Less

EINECSNumber : 219-505-4

TSCA : TSCA