Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

Alfa Aesar™ 2,4-Dichlorobenzenediazonium 1,5-naphthalenedisulfonate hydrate, 97%

Manufacturer:  Alfa Aesar™ H5572114

Quantity 25g

 View more versions of this product

Catalog No. AAH5572114

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

123333-91-5
MFCD08457580
2,4-Dichlorobenzenediazonium 1,5-naphthalenedisulfonate hydrate
461.29 (Anhydrous)
UN3261

Specifications

Hygroscopic
195°C (dec.)
123333-91-5
461.29 (Anhydrous)
UN3261
97%
Yellow to pale brown
25g
Powder
MFCD08457580
2,4-Dichlorobenzenediazonium 1,5-naphthalenedisulfonate hydrate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit