Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorobenzenesulfonyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
16271-33-3
C6H3Cl3O2S
245.498
MFCD00052712
FDTPBIKNYWQLAE-UHFFFAOYSA-N
2,4-dichloro-benzenesulfonyl chloride,2,4-dichlorobenzene-1-sulfonyl chloride,benzenesulfonyl chloride, 2,4-dichloro,2,4-dichlorobenzenesulphonyl chloride,2,4-dichlorobenzenesulfonylchloride,2,4-dichlorophenylsulfonyl chloride,2,4-dichlorobenzenesulphonylchloride,2,4-dichlorophenyl chlorosulfone,pubchem5115,acmc-1c9b8
204419
2,4-dichlorobenzenesulfonyl chloride
C1=CC(=C(C=C1Cl)Cl)S(=O)(=O)Cl
16271-33-3
C6H3Cl3O2S
245.498
MFCD00052712
FDTPBIKNYWQLAE-UHFFFAOYSA-N
2,4-dichloro-benzenesulfonyl chloride,2,4-dichlorobenzene-1-sulfonyl chloride,benzenesulfonyl chloride, 2,4-dichloro,2,4-dichlorobenzenesulphonyl chloride,2,4-dichlorobenzenesulfonylchloride,2,4-dichlorophenylsulfonyl chloride,2,4-dichlorobenzenesulphonylchloride,2,4-dichlorophenyl chlorosulfone,pubchem5115,acmc-1c9b8
204419
2,4-dichlorobenzenesulfonyl chloride
C1=CC(=C(C=C1Cl)Cl)S(=O)(=O)Cl
Color White to Beige
Melting Point 54°C to 58°C
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula Cl(Cl)C6H3(SO2Cl)
Formula Weight 245.51
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2, 4-Dichlorobenzenesulfonyl chloride
Show More Show Less

GHS P Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger