Alfa Aesar™ 2,4-Dichlorobenzoic acid, 98%

Manufacturer: Alfa Aesar™ A1237230

Quantity 250g

 View more versions of this product

Catalog No. AAA1237230

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 50-84-0
    Molecular Formula C7H4Cl2O2
    Formula Weight 191.01
    MDL Number MFCD00002414
    Beilstein 1868192
    Chemical Name or Material 2,4-Dichlorobenzoic acid

    Specifications

    Formula Weight 191.01
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Molecular Formula C7H4Cl2O2
    MDL Number MFCD00002414
    CAS 50-84-0
    Beilstein 1868192
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number DG6650000
    Melting Point 159° to 164°C
    Health Hazard 1 Warning
    Quantity 250g
    Chemical Name or Material 2,4-Dichlorobenzoic acid
    Assay 98%

    Description

    Applications
    2,4-Dichlorobenzoic acid is used as reagent during the synthesis of pyrimido[2 ft.,1 ft.:2,3]thiazolo[4,5-b]quinoxaline derivatives. It is also used as starting reagent during the synthesis of 1-(substituted)-1,4-dihydro-6-nitro-4-oxo-7-(sub-secondary amino)-quinoline-3-carboxylic acids.

    Solubility
    Soluble in ethanol (100 mg/ml), water (0.48 mg/ml), alcohol, ether, and acetone.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit