Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Dichlorobenzyl mercaptan, 98+%

Click to view available options
Quantity:
5g
25g
59293-67-3
C7H6Cl2S
193.085
MFCD00039650
ZSPXTTVUJDSRNJ-UHFFFAOYSA-N
2,4-dichlorobenzyl mercaptan,2,4-dichlorophenyl methanethiol,2,4-dichlorobenzylmercaptan,2,4-dichlorobenzenemethanethiol,2,4-dichlorotoluene-alpha-thiol,benzenemethanethiol, 2,4-dichloro,2,4-dichloro-alpha-toluenethiol,acmc-1b9kz,d0sz9f,2,4-dichlorothiobenzyl alcohol
95756
(2,4-dichlorophenyl)methanethiol
C1=CC(=C(C=C1Cl)Cl)CS
59293-67-3
C7H6Cl2S
193.085
MFCD00039650
ZSPXTTVUJDSRNJ-UHFFFAOYSA-N
2,4-dichlorobenzyl mercaptan,2,4-dichlorophenyl methanethiol,2,4-dichlorobenzylmercaptan,2,4-dichlorobenzenemethanethiol,2,4-dichlorotoluene-alpha-thiol,benzenemethanethiol, 2,4-dichloro,2,4-dichloro-alpha-toluenethiol,acmc-1b9kz,d0sz9f,2,4-dichlorothiobenzyl alcohol
95756
(2,4-dichlorophenyl)methanethiol
C1=CC(=C(C=C1Cl)Cl)CS
Flash Point >100°C (212°F)
Quantity 5g
Boiling Point 150°C to 151°C (12mmHg)
UN Number UN2810
Beilstein 509602
Formula Weight 193.1
Percent Purity ≥98%
Chemical Name or Material 2,4-Dichlorobenzyl mercaptan

GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.