Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(2,4-Dichlorobenzyl)triphenylphosphonium chloride hydrate, 98%, Acros Organics
Click to view available options
Quantity:
25g
Description
for C-C Bond Formation (Olefination)
Specifications
Quantity | 25g |
Color | White |
Infrared Spectrum | Authentic |
Water | 8% max. (K.F.) |
Formula Weight | 457.77 |
Physical Form | Crystalline Powder |
Percent Purity | ≥97.5% (on dry substance) |