Alfa Aesar™ 2,4-Dichlorobenzylideneacetone, 97%

Manufacturer: Alfa Aesar™ B2527006

Quantity 5g

 View more versions of this product

Catalog No. AAB2527006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 61888-78-6
    Molecular Formula C10H8Cl2O
    Formula Weight 215.08
    MDL Number MFCD00052852
    Synonym 4-(2,4-Dichlorophenyl)-3-buten-2-one
    Chemical Name or Material 2,4-Dichlorobenzylideneacetone

    Specifications

    Formula Weight 215.08
    Molecular Formula C10H8Cl2O
    Melting Point 83° to 85°C
    MDL Number MFCD00052852
    CAS 61888-78-6
    Synonym 4-(2,4-Dichlorophenyl)-3-buten-2-one
    Quantity 5g
    Chemical Name or Material 2,4-Dichlorobenzylideneacetone
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit