Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Dichlorobenzylideneacetone, 97%

Click to view available options
Quantity:
1g
5g
25g
61888-78-6
C10H8Cl2O
215.073
MFCD00052852
SAEQHTBHPNKKRX-NSCUHMNNSA-N
2,4-dichlorobenzylideneacetone,4-2,4-dichlorophenyl-3-buten-2-one,e-4-2,4-dichlorophenyl but-3-en-2-one,3e-4-2,4-dichlorophenyl but-3-en-2-one,e-4-2,4-dichloro-phenyl-but-3-en-2-one,4-2,4-dichlorophenyl-3-butene-2-one
5849647
(E)-4-(2,4-dichlorophenyl)but-3-en-2-one
CC(=O)C=CC1=C(C=C(C=C1)Cl)Cl
61888-78-6
C10H8Cl2O
215.073
MFCD00052852
SAEQHTBHPNKKRX-NSCUHMNNSA-N
2,4-dichlorobenzylideneacetone,4-2,4-dichlorophenyl-3-buten-2-one,e-4-2,4-dichlorophenyl but-3-en-2-one,3e-4-2,4-dichlorophenyl but-3-en-2-one,e-4-2,4-dichloro-phenyl-but-3-en-2-one,4-2,4-dichlorophenyl-3-butene-2-one
5849647
(E)-4-(2,4-dichlorophenyl)but-3-en-2-one
CC(=O)C=CC1=C(C=C(C=C1)Cl)Cl
Quantity 1g
Melting Point 83°C to 85°C
Formula Weight 215.08
Percent Purity 97%
Chemical Name or Material 2,4-Dichlorobenzylideneacetone