Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorophenethyl alcohol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
81156-68-5
C8H8Cl2O
191.06
MFCD00192010
JIJGKPVJAALUQQ-UHFFFAOYSA-N
2,4-dichlorophenethyl alcohol,2-2,4-dichlorophenyl ethanol,2,4-dichlorophenethylalcohol,2-2,4-dichlorophenyl ethan-1-ol,2,4-dichlorobenzeneethanol,acmc-209pku,ksc450o3l,2,4-dichlorophenylmethyl-carbinol,2-2,4-dichlorophenyl-ethanol
2734099
2-(2,4-dichlorophenyl)ethanol
C1=CC(=C(C=C1Cl)Cl)CCO
81156-68-5
C8H8Cl2O
191.06
MFCD00192010
JIJGKPVJAALUQQ-UHFFFAOYSA-N
2,4-dichlorophenethyl alcohol,2-2,4-dichlorophenyl ethanol,2,4-dichlorophenethylalcohol,2-2,4-dichlorophenyl ethan-1-ol,2,4-dichlorobenzeneethanol,acmc-209pku,ksc450o3l,2,4-dichlorophenylmethyl-carbinol,2-2,4-dichlorophenyl-ethanol
2734099
2-(2,4-dichlorophenyl)ethanol
C1=CC(=C(C=C1Cl)Cl)CCO
Boiling Point 119°C (0.8mmHg)
Specific Gravity 1.321
Flash Point >110°C
Refractive Index 1.5655 to 1.5675
Color Colorless
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Density 1.3210g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 191.06
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2, 4-Dichlorophenethyl alcohol
Show More Show Less