Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorophenethylamine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
52516-13-9
C8H9Cl2N
190.07
MFCD00060611
VHJKDOLGYMULOP-UHFFFAOYSA-N
2,4-dichlorophenethylamine,2-2,4-dichlorophenyl ethanamine,2-2,4-dichlorophenyl ethylamine,2-2,4-dichlorophenyl ethan-1-amine,benzeneethanamine, 2,4-dichloro,acmc-209kzz,2,4-dichlorophenethyl amine,ksc497e8j,benzeneethanamine,2,4-dichloro
142938
2-(2,4-dichlorophenyl)ethanamine
C1=CC(=C(C=C1Cl)Cl)CCN
52516-13-9
C8H9Cl2N
190.07
MFCD00060611
VHJKDOLGYMULOP-UHFFFAOYSA-N
2,4-dichlorophenethylamine,2-2,4-dichlorophenyl ethanamine,2-2,4-dichlorophenyl ethylamine,2-2,4-dichlorophenyl ethan-1-amine,benzeneethanamine, 2,4-dichloro,acmc-209kzz,2,4-dichlorophenethyl amine,ksc497e8j,benzeneethanamine,2,4-dichloro
142938
2-(2,4-dichlorophenyl)ethanamine
C1=CC(=C(C=C1Cl)Cl)CCN
Specific Gravity 1.26
Density 1.2600g/mL
Color Colorless to Yellow
Flash Point >110°C
Refractive Index 1.5645 to 1.5665
Packaging Glass bottle
Quantity 25g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula Cl2C6H3CH2CH2NH2
Beilstein 12,IV,2491
Formula Weight 190.07
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2, 4-Dichlorophenethylamine, 98%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

GHS Signal Word: Warning

TSCA : TSCA