Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Dichlorophenol, 99%

Manufacturer:  Alfa Aesar™ A1206522

Quantity 100g

 View more versions of this product

Catalog No. AAA1206522


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

120-83-2
163
UN2928
14,3072
C6H4Cl2O
MFCD00002169
742467
2,4-Dichlorophenol

Specifications

114°C (237°F)
40° to 44°C
120-83-2
Phenol-like
MFCD00002169
742467
2,4-Dichlorophenol
209° to 210°C
100g
C6H4Cl2O
163
UN2928
14,3072
99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H311-H314-H318-H302
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P280-P273-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.

RTECSNumber : SK8575000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit