Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorophenylacetic Acid 99%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
19719-28-9
C8H6Cl2O2
205.034
MFCD00004318
GXMWLJKTGBZMBH-UHFFFAOYSA-N
2,4-dichlorophenylacetic acid,2-2,4-dichlorophenyl acetic acid,benzeneacetic acid, 2,4-dichloro,2,4-dichloro-phenyl-acetic acid,2,4-dichlorophenyl acetic acid,2 ,4-dichlorophenylacetic acid,zlchem 477,pubchem15026,acmc-209f0x
88209
2-(2,4-dichlorophenyl)acetic acid
C1=CC(=C(C=C1Cl)Cl)CC(=O)O
19719-28-9
C8H6Cl2O2
205.034
MFCD00004318
GXMWLJKTGBZMBH-UHFFFAOYSA-N
2,4-dichlorophenylacetic acid,2-2,4-dichlorophenyl acetic acid,benzeneacetic acid, 2,4-dichloro,2,4-dichloro-phenyl-acetic acid,2,4-dichlorophenyl acetic acid,2 ,4-dichlorophenylacetic acid,zlchem 477,pubchem15026,acmc-209f0x
88209
2-(2,4-dichlorophenyl)acetic acid
C1=CC(=C(C=C1Cl)Cl)CC(=O)O
Quantity 50g
Packaging Glass bottle
Melting Point 128°C to 133°C
Color Beige-Yellow to White
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula Cl2C6H3CH2COOH
Beilstein 09, III, 2271
Formula Weight 205.04
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 2, 4-Dichlorophenylacetic acid, 99%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 243-248-7

TSCA : TSCA