Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorophenylacetonitrile, ≥99%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
6306-60-1
C8H5Cl2N
186.035
MFCD00001899
VJARIBGMDPJLCL-UHFFFAOYSA-N
2,4-dichlorophenylacetonitrile,2-2,4-dichlorophenyl acetonitrile,2,4-dichlorobenzyl cyanide,benzeneacetonitrile, 2,4-dichloro,2,4-dichlorophenyl acetonitrile,2,4-dichlorobenzeneacetonitrile,unii-d9j4c5m11e,acetonitrile, 2,4-dichlorophenyl,2,4-dichloro-phenyl-acetonitrile,2,4-dichlorphenyl acetonitril
80566
2-(2,4-dichlorophenyl)acetonitrile
C1=CC(=C(C=C1Cl)Cl)CC#N
6306-60-1
C8H5Cl2N
186.035
MFCD00001899
VJARIBGMDPJLCL-UHFFFAOYSA-N
2,4-dichlorophenylacetonitrile,2-2,4-dichlorophenyl acetonitrile,2,4-dichlorobenzyl cyanide,benzeneacetonitrile, 2,4-dichloro,2,4-dichlorophenyl acetonitrile,2,4-dichlorobenzeneacetonitrile,unii-d9j4c5m11e,acetonitrile, 2,4-dichlorophenyl,2,4-dichloro-phenyl-acetonitrile,2,4-dichlorphenyl acetonitril
80566
2-(2,4-dichlorophenyl)acetonitrile
C1=CC(=C(C=C1Cl)Cl)CC#N
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 60°C to 63°C
Quantity 25g
Color Yellow to Beige
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula Cl2C6H3CH2CN
Formula Weight 186.04
Physical Form Crystalline Solid
Percent Purity ≥99.0%
Chemical Name or Material 2, 4-Dichlorophenylacetonitrile, 99+%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 228-621-4

TSCA : TSCA