Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorophenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
68716-47-2
C6H5BCl2O2
190.82
MFCD00013930
QNEGDGPAXKYZHZ-UHFFFAOYSA-N
2,4-dichlorophenyl boronic acid,2,4-dichlorobenzeneboronic acid,2,4-dichlorophenylbornic acid,2,4-dichlorophenyl boranediol,chembl20726,2,4-dichloro benzene boronic acid,boronic acid, 2,4-dichlorophenyl,2,4-dichlorobenzeneboron dihydroxide,2,4-dichlorobenzeneboronicacid
2734659
(2,4-dichlorophenyl)boronic acid
B(C1=C(C=C(C=C1)Cl)Cl)(O)O
68716-47-2
C6H5BCl2O2
190.82
MFCD00013930
QNEGDGPAXKYZHZ-UHFFFAOYSA-N
2,4-dichlorophenyl boronic acid,2,4-dichlorobenzeneboronic acid,2,4-dichlorophenylbornic acid,2,4-dichlorophenyl boranediol,chembl20726,2,4-dichloro benzene boronic acid,boronic acid, 2,4-dichlorophenyl,2,4-dichlorobenzeneboron dihydroxide,2,4-dichlorobenzeneboronicacid
2734659
(2,4-dichlorophenyl)boronic acid
B(C1=C(C=C(C=C1)Cl)Cl)(O)O
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
Quantity 5g
Melting Point 240.0°C to 245.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 190.82
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 2, 4-Dichlorophenylboronic acid, 97%
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face prot

GHS Signal Word: Warning

Recommended Storage : Refrigerator +4°C