Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloropyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
100g
26452-80-2
C5H3Cl2N
147.99
MFCD00955618
TYPVHTOETJVYIV-UHFFFAOYSA-N
pyridine, 2,4-dichloro,2,4-dichloro pyridine,2,4-dichoropyridine,2, 4-dichloropyridine,2,4-dichloro-pyridine,2,4-dichlorpyridin,2,4-dichlorpyridine,2,4-dichloropyrdine,pubchem3531,acmc-1cnvt
33515
2,4-dichloropyridine
C1=CN=C(C=C1Cl)Cl
26452-80-2
C5H3Cl2N
147.99
MFCD00955618
TYPVHTOETJVYIV-UHFFFAOYSA-N
pyridine, 2,4-dichloro,2,4-dichloro pyridine,2,4-dichoropyridine,2, 4-dichloropyridine,2,4-dichloro-pyridine,2,4-dichlorpyridin,2,4-dichlorpyridine,2,4-dichloropyrdine,pubchem3531,acmc-1cnvt
33515
2,4-dichloropyridine
C1=CN=C(C=C1Cl)Cl
Specific Gravity 1.37
Flash Point 89°C
Quantity 1g
Density 1.3700g/mL
Melting Point -1.0°C
Refractive Index 1.5500 to 1.5540
Packaging Glass bottle
Boiling Point 189.0°C to 190.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 147.99
Percent Purity 98%
Chemical Name or Material 2, 4-Dichloropyridine
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse

GHS Signal Word: Warning

EINECSNumber : 247-717-7