Alfa Aesar™ 2,4-Difluoro-3-methylbenzoyl chloride, 98%

Manufacturer: Alfa Aesar™ H3165803

Quantity 1g

 View more versions of this product

Catalog No. AAH3165803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 112857-70-2
    Molecular Formula C8H5ClF2O
    Formula Weight 190.58
    MDL Number MFCD07777157
    UN Number UN3261
    Synonym 2,4-Difluoro-m-toluoyl chloride
    Chemical Name or Material 2,4-Difluoro-3-methylbenzoyl chloride

    Specifications

    Formula Weight 190.58
    Molecular Formula C8H5ClF2O
    MDL Number MFCD07777157
    CAS 112857-70-2
    UN Number UN3261
    Synonym 2,4-Difluoro-m-toluoyl chloride
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Melting Point 35° to 38°C
    Quantity 1g
    Chemical Name or Material 2,4-Difluoro-3-methylbenzoyl chloride
    Assay 98%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit