Alfa Aesar™ 2,4-Difluoro-3-methylbenzylamine, 98%

Manufacturer: Alfa Aesar™ H3206006

Quantity 5g

 View more versions of this product

Catalog No. AAH3206006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 847502-90-3
    Molecular Formula C8H9F2N
    Formula Weight 157.16
    MDL Number MFCD08275293
    UN Number UN2735
    Chemical Name or Material 2,4-Difluoro-3-methylbenzylamine

    Specifications

    Formula Weight 157.16
    MDL Number MFCD08275293
    CAS 847502-90-3
    UN Number UN2735
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    Boiling Point 42°C (1mmHg)
    Quantity 5g
    Molecular Formula C8H9F2N
    Recommended Storage 2° to 8°C
    Chemical Name or Material 2,4-Difluoro-3-methylbenzylamine
    Assay 98%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    2° to 8°C

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit