Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2',4'-Difluoro-N-methylacetanilide, 97%

Click to view available options
Quantity:
1g
5g
25g
238403-47-9
C9H9F2NO
185.174
MFCD01567301
XWGIBKPPTIGDDI-UHFFFAOYSA-N
n-2,4-difluorophenyl-n-methylacetamide,2',4'-difluoro-n-methylacetanilide,acmc-20akd8,maybridge3_007484,acetamide,n-2,4-difluorophenyl-n-methyl
2737628
N-(2,4-difluorophenyl)-N-methylacetamide
CC(=O)N(C)C1=C(C=C(C=C1)F)F
238403-47-9
C9H9F2NO
185.174
MFCD01567301
XWGIBKPPTIGDDI-UHFFFAOYSA-N
n-2,4-difluorophenyl-n-methylacetamide,2',4'-difluoro-n-methylacetanilide,acmc-20akd8,maybridge3_007484,acetamide,n-2,4-difluorophenyl-n-methyl
2737628
N-(2,4-difluorophenyl)-N-methylacetamide
CC(=O)N(C)C1=C(C=C(C=C1)F)F
Quantity 5g
Melting Point 83°C to 85°C
Formula Weight 185.17
Percent Purity 97%
Chemical Name or Material 2',4'-Difluoro-N-methylacetanilide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning