Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Difluoroanisole, 98%

Click to view available options
Quantity:
5g
25g
452-10-8
C7H6F2O
144.121
MFCD00010731
CRMJLJFDPNJIQA-UHFFFAOYSA-N
2,4-difluoroanisole,2,4-difluoroanilsole,2,4-difluoromethoxy benzene,benzene, 2,4-difluoro-1-methoxy,pubchem3455,2,4-difluoro anisole,ksc493m8b,2,4-difluoro-1-methoxy-benzene,#
136293
2,4-difluoro-1-methoxybenzene
COC1=C(C=C(C=C1)F)F
452-10-8
C7H6F2O
144.121
MFCD00010731
CRMJLJFDPNJIQA-UHFFFAOYSA-N
2,4-difluoroanisole,2,4-difluoroanilsole,2,4-difluoromethoxy benzene,benzene, 2,4-difluoro-1-methoxy,pubchem3455,2,4-difluoro anisole,ksc493m8b,2,4-difluoro-1-methoxy-benzene,#
136293
2,4-difluoro-1-methoxybenzene
COC1=C(C=C(C=C1)F)F
Boiling Point 53°C (18mmHg)
Flash Point 51°C (123°F)
Quantity 25g
UN Number UN1993
Beilstein 2518264
Formula Weight 144.12
Percent Purity 98%
Chemical Name or Material 2,4-Difluoroanisole

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.