Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Difluorobenzeneboronic acid, 97%

Click to view available options
Quantity:
1g
5g
25g
144025-03-6
C6H5BF2O2
157.911
MFCD01318998
QQLRSCZSKQTFGY-UHFFFAOYSA-N
2,4-difluorobenzeneboronic acid,2,4-difluorophenyl boronic acid,2,4-fluorobenzene boronic acid,2,4-difluorophenyl boranediol,2,4-difluorophenyl-boronic acid,2,4-difluorophenylboronicacid,2 4-difluorophenylboronic acid,boronic acid, 2,4-difluorophenyl,boronic acid, b-2,4-difluorophenyl
2734334
(2,4-difluorophenyl)boronic acid
B(C1=C(C=C(C=C1)F)F)(O)O
144025-03-6
C6H5BF2O2
157.911
MFCD01318998
QQLRSCZSKQTFGY-UHFFFAOYSA-N
2,4-difluorobenzeneboronic acid,2,4-difluorophenyl boronic acid,2,4-fluorobenzene boronic acid,2,4-difluorophenyl boranediol,2,4-difluorophenyl-boronic acid,2,4-difluorophenylboronicacid,2 4-difluorophenylboronic acid,boronic acid, 2,4-difluorophenyl,boronic acid, b-2,4-difluorophenyl
2734334
(2,4-difluorophenyl)boronic acid
B(C1=C(C=C(C=C1)F)F)(O)O
Melting Point 247°C to 250°C
Quantity 1g
Beilstein 8616257
Formula Weight 157.91
Percent Purity 97%
Chemical Name or Material 2,4-Difluorobenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning