Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Difluorobenzenesulfonyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
13918-92-8
C6H3ClF2O2S
212.6
MFCD00042481
FJSAJUXIHJIAMD-UHFFFAOYSA-N
2,4-difluorobenzene-1-sulfonyl chloride, 2,4-difluorobenzenesulphonyl chloride, 2,4-difluoro-benzenesulfonyl chloride, benzenesulfonyl chloride, 2,4-difluoro, 2,4-difluorophenyl chlorosulfone, buttpark 23\07-79, 2,4-difluorophenylsulfonyl chloride, pubchem5122, acmc-209civ
2734273
2,4-difluorobenzenesulfonyl chloride
C1=CC(=C(C=C1F)F)S(=O)(=O)Cl
13918-92-8
C6H3ClF2O2S
212.6
MFCD00042481
FJSAJUXIHJIAMD-UHFFFAOYSA-N
2,4-difluorobenzene-1-sulfonyl chloride, 2,4-difluorobenzenesulphonyl chloride, 2,4-difluoro-benzenesulfonyl chloride, benzenesulfonyl chloride, 2,4-difluoro, 2,4-difluorophenyl chlorosulfone, buttpark 23\07-79, 2,4-difluorophenylsulfonyl chloride, pubchem5122, acmc-209civ
2734273
2,4-difluorobenzenesulfonyl chloride
C1=CC(=C(C=C1F)F)S(=O)(=O)Cl
Specific Gravity 1.581
Boiling Point 101°C to 102°C (1.0mmHg)
Refractive Index 1.5187
Density 1.5810g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 1g
Color Colorless to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (ex Cl) (Argentometry)
Linear Formula (F)2C6H3SO2Cl
Formula Weight 212.6
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2, 4-Difluorobenzenesulfonyl chloride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger