Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Difluorobenzophenone, 97+%

Click to view available options
Quantity:
5g
25g
85068-35-5
C13H8F2O
218.203
MFCD00000331
FRHMSTHFVFIPCO-UHFFFAOYSA-N
2,4-difluorobenzophenone,2,4-difluorophenyl phenyl methanone,2,4-difluorophenyl-phenylmethanone,methanone, 2,4-difluorophenyl phenyl,2,4-difluorophenyl phenyl ketone,fr cf dvr,2,4-bis fluoranyl phenyl-phenyl-methanone,2,4-difluorobenzo-phenone,frhmsthfvfipco-uhfffaoysa,2,4-difluorophenyl-phenyl-methanone
570834
(2,4-difluorophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=C(C=C(C=C2)F)F
85068-35-5
C13H8F2O
218.203
MFCD00000331
FRHMSTHFVFIPCO-UHFFFAOYSA-N
2,4-difluorobenzophenone,2,4-difluorophenyl phenyl methanone,2,4-difluorophenyl-phenylmethanone,methanone, 2,4-difluorophenyl phenyl,2,4-difluorophenyl phenyl ketone,fr cf dvr,2,4-bis fluoranyl phenyl-phenyl-methanone,2,4-difluorobenzo-phenone,frhmsthfvfipco-uhfffaoysa,2,4-difluorophenyl-phenyl-methanone
570834
(2,4-difluorophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=C(C=C(C=C2)F)F
Quantity 25g
Boiling Point 119°C to 120°C (2mmHg)
Flash Point >110°C (230°F)
Beilstein 6596192
Formula Weight 218.2
Percent Purity ≥97%
Chemical Name or Material 2,4-Difluorobenzophenone

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning