Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Difluoronitrobenzene, 99%, ACROS Organics™

Click to view available options
Quantity:
25mL
Packaging:
Glass bottle
446-35-5
C6H3F2NO2
159.092
MFCD00007050
RJXOVESYJFXCGI-UHFFFAOYSA-N
2,4-difluoronitrobenzene,benzene, 2,4-difluoro-1-nitro,1-nitro-2,4-difluorobenzene,1,3-difluoro-4-nitrobenzene,unii-6398y1d7js,2,4-difluoro-1-nitro-benzene,pubchem2288,2,4-difluornitrobenzol,2,4-difluoronitrobenzol,acmc-1akft
67967
2,4-difluoro-1-nitrobenzene
C1=CC(=C(C=C1F)F)[N+](=O)[O-]
446-35-5
C6H3F2NO2
159.092
MFCD00007050
RJXOVESYJFXCGI-UHFFFAOYSA-N
2,4-difluoronitrobenzene,benzene, 2,4-difluoro-1-nitro,1-nitro-2,4-difluorobenzene,1,3-difluoro-4-nitrobenzene,unii-6398y1d7js,2,4-difluoro-1-nitro-benzene,pubchem2288,2,4-difluornitrobenzol,2,4-difluoronitrobenzol,acmc-1akft
67967
2,4-difluoro-1-nitrobenzene
C1=CC(=C(C=C1F)F)[N+](=O)[O-]
Color Brown to Yellow
Quantity 25mL
Refractive Index 1.5110 to 1.5130 (20°C, 589nm)
Density 1.4500g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula F2C6H3NO2
Beilstein 05, I, 129
Formula Weight 159.09
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 2, 4-Difluoronitrobenzene
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger