Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Difluorophenylacetone, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
274682-91-6
C9H8F2O
170.16
MFCD02258866
XVHHDTNIOCSTLH-UHFFFAOYSA-N
1-2,4-difluorophenyl propan-2-one,2,4-difluorophenylacetone,2-propanone, 1-2,4-difluorophenyl,2,4-difluorophenyl propanone,1-2,4-difluorophenyl-2-propanone,1-2,4-difluoro-phenyl-propane-2-one
2734881
1-(2,4-difluorophenyl)propan-2-one
CC(=O)CC1=C(C=C(C=C1)F)F
274682-91-6
C9H8F2O
170.16
MFCD02258866
XVHHDTNIOCSTLH-UHFFFAOYSA-N
1-2,4-difluorophenyl propan-2-one,2,4-difluorophenylacetone,2-propanone, 1-2,4-difluorophenyl,2,4-difluorophenyl propanone,1-2,4-difluorophenyl-2-propanone,1-2,4-difluoro-phenyl-propane-2-one
2734881
1-(2,4-difluorophenyl)propan-2-one
CC(=O)CC1=C(C=C(C=C1)F)F
Packaging Glass bottle
Color Yellow
Infrared Spectrum Authentic
Quantity 1g
Refractive Index 1.4770 to 1.4790
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (F)2C6H3CH2COCH3
Formula Weight 170.16
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2, 4-Difluorophenylacetone
Show More Show Less