Alfa Aesar™ 2',4'-Difluoropropiophenone, 98%

Manufacturer: Alfa Aesar™ A1203222

Quantity 100g

 View more versions of this product

Catalog No. AAA1203222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 85068-30-0
    Molecular Formula C9H8F2O
    Formula Weight 170.16
    MDL Number MFCD00015507
    Beilstein 7633370
    Chemical Name or Material 2',4'-Difluoropropiophenone

    Specifications

    Formula Weight 170.16
    Molecular Formula C9H8F2O
    MDL Number MFCD00015507
    CAS 85068-30-0
    Boiling Point 51° to 52°C (1.5mmHg)
    Beilstein 7633370
    Flash Point 76°C (168°F)
    Quantity 100g
    Chemical Name or Material 2',4'-Difluoropropiophenone
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit