Alfa Aesar™ 2,4-Difluorothiophenol, 97%

Manufacturer: Alfa Aesar™ B2472614

Quantity 25g

 View more versions of this product

Catalog No. AAB2472614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1996-44-7
    Molecular Formula C6H4F2S
    Formula Weight 146.16
    MDL Number MFCD00203477
    UN Number UN1228
    Beilstein 2516728
    Synonym 2,4-Difluorobenzenethiol
    Chemical Name or Material 2,4-Difluorothiophenol

    Specifications

    Formula Weight 146.16
    Molecular Formula C6H4F2S
    MDL Number MFCD00203477
    UN Number UN1228
    Beilstein 2516728
    Synonym 2,4-Difluorobenzenethiol
    Health Hazard 2 GHS H Statement
    H331-H226-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Flammable liquid and vapour.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 53°C (127°F)
    Boiling Point 52°C (15mmHg)
    Quantity 25g
    CAS 1996-44-7
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: MERCAPTANS, LIQUID, FLAMMABLE, TOXIC, N.O.S.
    Chemical Name or Material 2,4-Difluorothiophenol
    Assay 97%

    GHS H Statement
    H331-H226-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Flammable liquid and vapour.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit