Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2',4'-Dihydroxy-2-(4-hydroxyphenyl)acetophenone, 97%

Click to view available options
Quantity:
250mg
1g
17720-60-4
C14H12O4
244.246
MFCD00498162
KLFCJXAPIFIIFR-UHFFFAOYSA-N
1-2,4-dihydroxyphenyl-2-4-hydroxyphenyl ethanone,2,4,4'-trihydroxydeoxybenzoin,2,4,4'-trihydroxy deoxybenzoin,1-2,4-dihydroxy-phenyl-2-4-hydroxy-phenyl-ethanone,1-2,4-dihydroxy-phenyl 2 4-hydro xy-phenyl-ethanone,ethanone,1-2,4-dihydroxyphenyl-2-4-hydroxyphenyl,ethanone, 1-2,4-dihydroxyphenyl-2-4-hydroxyphenyl,2,4,4'-terthydroxydeoxybenzoin,2,4,4-trihydroxydeoxybenzoin,2,4,4/'-trihydroxydeoxybenzoin
640096
1-(2,4-dihydroxyphenyl)-2-(4-hydroxyphenyl)ethanone
C1=CC(=CC=C1CC(=O)C2=C(C=C(C=C2)O)O)O
17720-60-4
C14H12O4
244.246
MFCD00498162
KLFCJXAPIFIIFR-UHFFFAOYSA-N
1-2,4-dihydroxyphenyl-2-4-hydroxyphenyl ethanone,2,4,4'-trihydroxydeoxybenzoin,2,4,4'-trihydroxy deoxybenzoin,1-2,4-dihydroxy-phenyl-2-4-hydroxy-phenyl-ethanone,1-2,4-dihydroxy-phenyl 2 4-hydro xy-phenyl-ethanone,ethanone,1-2,4-dihydroxyphenyl-2-4-hydroxyphenyl,ethanone, 1-2,4-dihydroxyphenyl-2-4-hydroxyphenyl,2,4,4'-terthydroxydeoxybenzoin,2,4,4-trihydroxydeoxybenzoin,2,4,4/'-trihydroxydeoxybenzoin
640096
1-(2,4-dihydroxyphenyl)-2-(4-hydroxyphenyl)ethanone
C1=CC(=CC=C1CC(=O)C2=C(C=C(C=C2)O)O)O
Quantity 250mg
Beilstein 1288279
Formula Weight 244.25
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2',4'-Dihydroxy-2-(4-hydroxyphenyl)acetophenone