Alfa Aesar™ 2,4-Dihydroxy-6-methylbenzoic acid hydrate, 97%

Manufacturer: Alfa Aesar™ H2755606

Quantity 5g

 View more versions of this product

Catalog No. AAH2755606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 480-64-8
    Molecular Formula C8H8O4·xH2O
    Formula Weight 168.15 (Anhydrous)
    MDL Number MFCD00210536
    Chemical Name or Material 2,4-Dihydroxy-6-methylbenzoic acid hydrate

    Specifications

    Formula Weight 168.15 (Anhydrous)
    Quantity 5g
    Molecular Formula C8H8O4·xH2O
    MDL Number MFCD00210536
    CAS 480-64-8
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point ≈175°C
    Chemical Name or Material 2,4-Dihydroxy-6-methylbenzoic acid hydrate
    Assay 97%

    Description

    Applications
    Orsellinic acid is a benzoic acid that has been shown to block PAF-mediated neuronal apoptosis. Orsellinic acid also shows free radical scavenging activity.

    Solubility
    Soluble in water, dimethyl sulfoxide or 100% ethanol.

    Notes
    Store in cool dry conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit