Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-dihydroxy-6-methylpyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
3749-51-7
C6H7NO2
125.13
MFCD01860849
WKGSLYHMRQRARV-UHFFFAOYSA-N
6-methylpyridine-2,4-diol,2,4-dihydroxy-6-methylpyridine,4-hydroxy-6-methylpyridin-2 1h-one,4-hydroxy-6-methyl-2 1h-pyridinone,2 1h-pyridinone, 4-hydroxy-6-methyl,6-methyl-2,4-pyridinediol,2-hydroxy-6-methyl-1h-pyridin-4-one,2,4-dihydroxy-6-methyl-pyridine,4-hydroxy-6-methyl-2-pyridinone,2 1h-pyridone, 4-hydroxy-6-methyl
54691419
4-hydroxy-6-methyl-1H-pyridin-2-one
CC1=CC(=CC(=O)N1)O
3749-51-7
C6H7NO2
125.13
MFCD01860849
WKGSLYHMRQRARV-UHFFFAOYSA-N
6-methylpyridine-2,4-diol,2,4-dihydroxy-6-methylpyridine,4-hydroxy-6-methylpyridin-2 1h-one,4-hydroxy-6-methyl-2 1h-pyridinone,2 1h-pyridinone, 4-hydroxy-6-methyl,6-methyl-2,4-pyridinediol,2-hydroxy-6-methyl-1h-pyridin-4-one,2,4-dihydroxy-6-methyl-pyridine,4-hydroxy-6-methyl-2-pyridinone,2 1h-pyridone, 4-hydroxy-6-methyl
54691419
4-hydroxy-6-methyl-1H-pyridin-2-one
CC1=CC(=CC(=O)N1)O
Packaging Glass bottle
Infrared Spectrum Authentic
Color Beige
Quantity 1g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 125.13
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 2, 4-Dihydroxy-6-methylpyridine
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning