Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dihydroxy-6-methylpyrimidine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
Packaging:
Glass bottle
Plastic Bottle
626-48-2
C5H6N2O2
126.11
MFCD00006028
SHVCSCWHWMSGTE-UHFFFAOYSA-N
6-methyluracil,2,4-dihydroxy-6-methylpyrimidine,pseudothymine,6-methylpyrimidine-2,4 1h,3h-dione,4-methyluracil,6-methylpyrimidine-2,4-diol,2,4 1h,3h-pyrimidinedione, 6-methyl,uracil, 6-methyl,6-methyl-2,4 1h,3h-pyrimidinedione,2,4-pyrimidinediol, 6-methyl
12283
CHEBI:74733
6-methyl-1H-pyrimidine-2,4-dione
CC1=CC(=O)NC(=O)N1
626-48-2
C5H6N2O2
126.11
MFCD00006028
SHVCSCWHWMSGTE-UHFFFAOYSA-N
6-methyluracil,2,4-dihydroxy-6-methylpyrimidine,pseudothymine,6-methylpyrimidine-2,4 1h,3h-dione,4-methyluracil,6-methylpyrimidine-2,4-diol,2,4 1h,3h-pyrimidinedione, 6-methyl,uracil, 6-methyl,6-methyl-2,4 1h,3h-pyrimidinedione,2,4-pyrimidinediol, 6-methyl
12283
CHEBI:74733
6-methyl-1H-pyrimidine-2,4-dione
CC1=CC(=O)NC(=O)N1
Packaging Glass bottle
Melting Point 318.0°C
Infrared Spectrum Authentic
Color White
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 24, 342
Merck Index 15, 6209
Solubility Information Solubility in water: 7g/L water (22°C). Other solubilities: soluble in alkalies,insoluble in ethanol and ether
Formula Weight 126.11
Physical Form Fine Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2, 4-Dihydroxy-6-methylpyrimidine, 97%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of damaging fertility.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 210-949-4

RTECSNumber : YR0700000

TSCA : TSCA