Learn More
Medchemexpress LLC 2,4-dihydroxy-N'-(2-hydroxy-5-nitrobenzylidene)benzohydrazide | 304481-72-9 | 99.9% | 317.25 g·mol⁻1 | C14H11N3O6 | 5MG

Supplier: Medchemexpress LLC HY1232695MG
PKUMDL-WQ-2101 is a non-NAD+-competing allosteric inhibitor of phosphoglycerate dehydrogenase (PHGDH) with a reported IC50 of 34.8 μM. The compound exhibits antitumor activity in cellular models and is used to probe PHGDH function and serine biosynthesis. It is supplied as a solid in multiple small pack sizes and requires low-temperature storage and careful solvent handling.
- Allosteric PHGDH inhibitor with reported IC50 of 34.8 μM.
- High purity (~99.9%) suitable for biochemical and cellular studies.
- Molecular weight 317.25 g·mol⁻1; formula C14H11N3O6.
- Available in small pack sizes for research use.
- Storage: powder -20°C; in solvent -80°C (up to 6 months) or -20°C (up to 1 month).
- Handling: prepare aliquots and use newly opened anhydrous DMSO to avoid solubility issues and freeze-thaw inactivation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.