Alfa Aesar™ 2',4'-Dihydroxyacetophenone, 98%

Manufacturer: Alfa Aesar™ A1206618

Quantity 50g

 View more versions of this product

Catalog No. AAA1206618

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 89-84-9
    Molecular Formula C8H8O3
    Formula Weight 152.15
    MDL Number MFCD00002279
    Beilstein 1282505
    Merck Index 14,8140
    Synonym 4-Acetylresorcinol; Resacetophenone
    Chemical Name or Material 2',4'-Dihydroxyacetophenone

    Specifications

    Formula Weight 152.15
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 89-84-9
    Health Hazard 1 Danger
    RTECS Number AM7525000
    Quantity 50g
    Merck Index 14,8140
    Health Hazard 2 GHS H Statement
    H318-H335-H315
    Causes serious eye damage.
    May cause respiratory irritation.
    Causes skin irritation.
    TSCA TSCA
    Molecular Formula C8H8O3
    MDL Number MFCD00002279
    Melting Point 142° to 146°C
    Beilstein 1282505
    Synonym 4-Acetylresorcinol; Resacetophenone
    Chemical Name or Material 2',4'-Dihydroxyacetophenone
    Assay 98%

    GHS H Statement
    H318-H335-H315
    Causes serious eye damage.
    May cause respiratory irritation.
    Causes skin irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit