Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dihydroxybenzaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
95-01-2
C7H6O3
138.12
IUNJCFABHJZSKB-UHFFFAOYSA-N
beta-resorcylaldehyde,4-hydroxysalicylaldehyde,4-formylresorcinol,beta-resorcaldehyde,benzaldehyde, 2,4-dihydroxy,beta-resorcinaldehyde,beta-resorcylic aldehyde,2,4-dihydroxybenzenecarbonal,salicylaldehyde, 4-hydroxy,4-hydroxysalicyladehyde
7213
CHEBI:50198
2,4-dihydroxybenzaldehyde
C1=CC(=C(C=C1O)O)C=O
95-01-2
C7H6O3
138.12
IUNJCFABHJZSKB-UHFFFAOYSA-N
beta-resorcylaldehyde,4-hydroxysalicylaldehyde,4-formylresorcinol,beta-resorcaldehyde,benzaldehyde, 2,4-dihydroxy,beta-resorcinaldehyde,beta-resorcylic aldehyde,2,4-dihydroxybenzenecarbonal,salicylaldehyde, 4-hydroxy,4-hydroxysalicyladehyde
7213
CHEBI:50198
2,4-dihydroxybenzaldehyde
C1=CC(=C(C=C1O)O)C=O
Infrared Spectrum Authentic
Quantity 100g
Boiling Point 220.0°C to 228.0°C (22.0mmHg)
Melting Point 133.0°C to 138.0°C
Packaging Glass bottle
Color White to Beige
Assay Percent Range 97.5 % min. (GC)
Linear Formula (HO)2C6H3CHO
Beilstein 08,241
Merck Index 15,8277
Solubility Information Solubility in water: freely soluble. Other solubilities: freely soluble in alcohol,ether,chloroform and,glacial acetic acic
Formula Weight 138.12
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2, 4-Dihydroxybenzaldehyde
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 202-383-1