Learn More
Tocris Bioscience™ 2,4-Dihydroxyphenylacetyl-L-asparagine
Component of Joro spider toxin
Manufacturer:  Tocris Bioscience™ 0262/50
 View more versions of this product
Catalog No. 02-625-0
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Constituent of various spider toxins. Reported to be specific blocker of glutamate receptors.
Specifications
Specifications
111872-98-1
Observed MW: 282.25
Soluble to 50 mM in water
C12H14N2O6
50mg
Safety and Handling
Safety and Handling
Recommended Storage : Desiccate at -20°C